Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331331.1 | NP_001321462.1 | 4 - 5 | 1019 | 77 | |
NM_100112.3 | NP_171733.2 | 4 - 5 | 884 | 77 | ||
GenBank mRNA | AY081352.1 | - | 4 - 5 | 884 | 77 | |
BT002154.1 | - | 4 - 5 | 840 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |