Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331332.1 | NP_001323315.1 | 3 - 4 | 888 | 95 | |
NM_100115.3 | NP_563650.1 | 4 - 5 | 764 | 95 | ||
GenBank mRNA | AF288287.1 | - | 3 - 4 | 569 | 95 | |
AF323182.1 | - | 4 - 5 | 728 | 95 | ||
AF324245.1 | - | 4 - 5 | 643 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |