Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100139.4 | NP_563658.1 | 16 - 17 | 1957 | 69 | |
GenBank mRNA | AF060485.1 | - | 16 - 17 | 1907 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |