Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331364.1 | NP_001320623.1 | 6 - 7 | 1334 | 79 | |
NM_100172.3 | NP_171790.1 | 7 - 8 | 1278 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |