Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WEE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100178.3 | NP_171796.1 | 2 - 3 | 463 | 97 | |
GenBank mRNA | AB077385.1 | - | 2 - 3 | 385 | 97 | |
AJ439593.1 | - | 2 - 3 | 463 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |