Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083983.1 | NP_001077452.1 | 3 - 4 | 1101 | 79 | |
NM_001331388.1 | NP_001322638.1 | 3 - 4 | 1122 | 79 | ||
NM_001331389.1 | NP_001322639.1 | 3 - 4 | 1101 | 79 | ||
NM_100198.5 | NP_171815.3 | 3 - 4 | 1122 | 79 | ||
GenBank mRNA | AK176297.1 | - | 3 - 4 | 1094 | 79 | |
AK227660.1 | - | 3 - 4 | 1094 | 79 | ||
DQ462573.1 | - | 3 - 4 | 1101 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |