Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100225.3 | NP_563684.1 | 5 - 6 | 783 | 139 | |
GenBank mRNA | AB041767.1 | - | 5 - 6 | 427 | 139 | |
AK176256.1 | - | 5 - 6 | 702 | 139 | ||
AK176767.1 | - | 5 - 6 | 663 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |