Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RING1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197972.1 | NP_001184901.1 | 5 - 6 | 730 | 141 | |
NM_001331438.1 | NP_001318915.1 | 5 - 6 | 725 | 141 | ||
NM_001331439.1 | NP_001323369.1 | 4 - 5 | 1028 | 141 | ||
NM_001331440.1 | NP_001323368.1 | 4 - 5 | 950 | 141 | ||
NM_001331441.1 | NP_001323367.1 | 4 - 5 | 447 | 141 | ||
NM_100256.3 | NP_171873.2 | 5 - 6 | 705 | 141 | ||
GenBank mRNA | AK226183.1 | - | 5 - 6 | 707 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |