Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197977.1 | NP_001184906.1 | 8 - 9 | 4282 | 70 | |
NM_001331463.1 | NP_001322077.1 | 9 - 10 | 4149 | 70 | ||
NM_100293.3 | NP_171908.1 | 8 - 9 | 4315 | 70 | ||
GenBank mRNA | AF225908.1 | - | 8 - 9 | 4050 | 70 | |
Y11250.1 | - | 4030 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |