Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100339.2 | NP_171954.1 | 35 - 36 | 4994 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |