Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100351.5 | NP_171965.1 | 11 - 12 | 1118 | 97 | |
GenBank mRNA | AY099741.1 | - | 11 - 12 | 1094 | 97 | |
BT003415.1 | - | 11 - 12 | 1062 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |