Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100351.5 | NP_171965.1 | 4 - 5 | 413 | 59 | |
GenBank mRNA | AY099741.1 | - | 4 - 5 | 389 | 59 | |
BT003415.1 | - | 4 - 5 | 357 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |