Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331495.1 | NP_001322020.1 | 6 - 7 | 1247 | 120 | |
NM_001331496.1 | NP_001322021.1 | 4 - 5 | 719 | 120 | ||
NM_001331497.1 | NP_001322022.1 | 6 - 7 | 1270 | 120 | ||
NM_100352.3 | NP_171966.2 | 6 - 7 | 1287 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |