Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083995.4 | NP_001077464.4 | 9 - 10 | 1808 | 65 | |
NM_001331587.1 | NP_001321563.1 | 9 - 10 | 1922 | 65 | ||
NM_001331588.1 | NP_001321564.1 | 9 - 10 | 1922 | 65 | ||
NM_100464.7 | NP_172074.6 | 9 - 10 | 1892 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |