Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331589.1 | NP_001321426.1 | 2 - 3 | 995 | 65 | |
NM_100466.5 | NP_172076.1 | 2 - 3 | 710 | 65 | ||
GenBank mRNA | AF332458.1 | - | 1 - 2 | 386 | 65 | |
AF422178.1 | - | 1 - 2 | 420 | 65 | ||
AY034935.1 | - | 2 - 3 | 536 | 65 | ||
AY113925.1 | - | 1 - 2 | 386 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |