Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331643.1 | NP_001321998.1 | 3 - 4 | 759 | 84 | |
NM_100544.3 | NP_172152.1 | 3 - 4 | 782 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |