Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSBP-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123767.1 | NP_001117239.1 | 3 - 4 | 594 | 63 | |
NM_100545.7 | NP_172153.1 | 3 - 4 | 842 | 63 | ||
GenBank mRNA | AY056416.1 | - | 3 - 4 | 591 | 63 | |
AY070469.1 | - | 3 - 4 | 591 | 63 | ||
AY074308.1 | - | 3 - 4 | 593 | 63 | ||
AY087305.1 | - | 3 - 4 | 598 | 63 | ||
AY096487.1 | - | 3 - 4 | 536 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |