Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXO70B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100573.4 | NP_172181.1 | 1 - 2 | 726 | 81 | |
GenBank mRNA | AY075660.1 | - | 1 - 2 | 500 | 81 | |
AY101526.1 | - | 1 - 2 | 490 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |