Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112954.4 | NP_566666.2 | 17 - 18 | 2188 | 75 | |
GenBank mRNA | AK226894.1 | - | 17 - 18 | 2038 | 75 | |
AX765820.1 | - | 17 - 18 | 1981 | 75 | ||
AY140096.1 | - | 17 - 18 | 2043 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |