Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112954.4 | NP_566666.2 | 18 - 19 | 2292 | 61 | |
GenBank mRNA | AK226894.1 | - | 18 - 19 | 2142 | 61 | |
AX765820.1 | - | 18 - 19 | 2085 | 61 | ||
AY140096.1 | - | 18 - 19 | 2147 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |