Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112954.4 | NP_566666.2 | 5 - 6 | 1020 | 111 | |
GenBank mRNA | AK226894.1 | - | 5 - 6 | 870 | 111 | |
AX765820.1 | - | 5 - 6 | 813 | 111 | ||
AY140096.1 | - | 5 - 6 | 875 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |