Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112954.4 | NP_566666.2 | 7 - 8 | 1207 | 90 | |
GenBank mRNA | AK226894.1 | - | 7 - 8 | 1057 | 90 | |
AX765820.1 | - | 7 - 8 | 1000 | 90 | ||
AY140096.1 | - | 7 - 8 | 1062 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |