Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112969.3 | NP_188714.2 | 16 - 17 | 1852 | 93 | |
GenBank mRNA | AJ297843.1 | - | 16 - 17 | 1674 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |