Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112969.3 | NP_188714.2 | 7 - 8 | 845 | 73 | |
GenBank mRNA | AJ297843.1 | - | 7 - 8 | 667 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |