Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112975.3 | NP_188720.2 | 6 - 7 | 1133 | 69 | |
GenBank mRNA | AY506549.1 | - | 6 - 7 | 902 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |