Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NF-YA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112983.5 | NP_566670.1 | 4 - 5 | 575 | 79 | |
GenBank mRNA | AF412056.1 | - | 4 - 5 | 577 | 79 | |
AY087696.1 | - | 4 - 5 | 579 | 79 | ||
AY093971.1 | - | 3 - 4 | 435 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |