Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSBTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338498.1 | NP_001319602.1 | 1 | 422 | 57 | |
NM_112999.4 | NP_566674.2 | 1 | 462 | 57 | ||
GenBank mRNA | AF324686.1 | - | 1 | 349 | 57 | |
AF326859.1 | - | 1 | 348 | 57 | ||
AF339682.1 | - | 1 | 308 | 57 | ||
AY062605.1 | - | 1 | 350 | 57 | ||
AY093364.1 | - | 1 | 308 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |