Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125053.1 | NP_001118525.1 | 7 - 8 | 1151 | 91 | |
NM_001337088.1 | NP_001324585.1 | 8 - 9 | 1016 | 91 | ||
NM_001337089.1 | NP_001324587.1 | 8 - 9 | 1016 | 91 | ||
NM_001337090.1 | NP_001324586.1 | 3 - 4 | 630 | 91 | ||
NM_130011.3 | NP_181976.1 | 8 - 9 | 955 | 91 | ||
GenBank mRNA | AY074629.1 | - | 8 - 9 | 926 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |