Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130033.3 | NP_566024.1 | 5 - 6 | 619 | 81 | |
GenBank mRNA | AF156896.1 | - | 5 - 6 | 476 | 81 | |
BT025645.1 | - | 5 - 6 | 464 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |