Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202819.1 | NP_001189748.1 | 3 - 4 | 870 | 76 | |
NM_130083.5 | NP_182045.1 | 3 - 4 | 723 | 76 | ||
GenBank mRNA | AF357529.1 | - | 3 - 4 | 472 | 76 | |
BT024623.1 | - | 3 - 4 | 417 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |