Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPFTSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130140.3 | NP_566056.1 | 4 - 5 | 707 | 88 | |
GenBank mRNA | AF360125.1 | - | 4 - 5 | 577 | 88 | |
AJ010820.1 | - | 4 - 5 | 557 | 88 | ||
AY051026.1 | - | 4 - 5 | 547 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |