Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPFTSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130140.3 | NP_566056.1 | 8 - 9 | 1009 | 67 | |
GenBank mRNA | AF360125.1 | - | 8 - 9 | 879 | 67 | |
AJ010820.1 | - | 8 - 9 | 859 | 67 | ||
AY051026.1 | - | 8 - 9 | 849 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |