Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130142.4 | NP_182103.1 | 6 - 7 | 653 | 81 | |
GenBank mRNA | AY050806.1 | - | 6 - 7 | 540 | 81 | |
AY088930.1 | - | 6 - 7 | 549 | 81 | ||
AY113964.1 | - | 6 - 7 | 452 | 81 | ||
DQ442991.1 | - | 6 - 7 | 452 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |