Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130142.4 | NP_182103.1 | 8 - 9 | 835 | 113 | |
GenBank mRNA | AY050806.1 | - | 8 - 9 | 722 | 113 | |
AY088930.1 | - | 8 - 9 | 731 | 113 | ||
AY113964.1 | - | 8 - 9 | 634 | 113 | ||
DQ442991.1 | - | 8 - 9 | 634 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |