Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CB5-C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130230.3 | NP_182188.1 | 2 - 3 | 468 | 75 | |
GenBank mRNA | AK175189.1 | - | 2 - 3 | 296 | 75 | |
AY087658.1 | - | 2 - 3 | 296 | 75 | ||
BT025600.1 | - | 2 - 3 | 142 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |