Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSI-P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036474.4 | NP_001031551.1 | 2 - 3 | 354 | 61 | |
NM_130248.5 | NP_566086.1 | 1 - 2 | 543 | 61 | ||
GenBank mRNA | AY065156.1 | - | 1 - 2 | 299 | 61 | |
AY114583.1 | - | 1 - 2 | 162 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |