Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036478.2 | NP_001031555.1 | 2 - 3 | 318 | 68 | |
NM_001084605.1 | NP_001078074.1 | 2 - 3 | 318 | 68 | ||
NM_130315.5 | NP_182269.1 | 2 - 3 | 484 | 68 | ||
NM_201979.2 | NP_973708.1 | 2 - 3 | 318 | 68 | ||
GenBank mRNA | AF083688.1 | - | 2 - 3 | 272 | 68 | |
AJ271470.1 | - | 2 - 3 | 476 | 68 | ||
AK316899.2 | - | 2 - 3 | 304 | 68 | ||
AY074348.1 | - | 2 - 3 | 317 | 68 | ||
AY091388.1 | - | 2 - 3 | 235 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |