Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MHX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130328.3 | NP_566105.1 | 2 - 3 | 722 | 79 | |
GenBank mRNA | AF109178.1 | - | 2 - 3 | 555 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |