Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130365.4 | NP_566119.1 | 10 - 11 | 1627 | 126 | |
GenBank mRNA | AJ242965.1 | - | 10 - 11 | 1501 | 126 | |
AY063915.1 | - | 10 - 11 | 1526 | 126 | ||
AY091270.1 | - | 10 - 11 | 1411 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |