Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331741.1 | NP_001318950.1 | 21 - 22 | 2793 | 139 | |
NM_100665.3 | NP_172269.2 | 21 - 22 | 2671 | 139 | ||
GenBank mRNA | AK230326.1 | - | 21 - 22 | 2638 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |