Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100683.3 | NP_172287.1 | 6 - 7 | 813 | 71 | |
GenBank mRNA | AY122907.1 | - | 6 - 7 | 748 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |