Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100689.5 | NP_172293.2 | 13 - 14 | 2215 | 67 | |
GenBank mRNA | AK117238.1 | - | 13 - 14 | 2223 | 67 | |
BT005964.1 | - | 13 - 14 | 2084 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |