Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100689.5 | NP_172293.2 | 1 - 2 | 915 | 75 | |
GenBank mRNA | AK117238.1 | - | 1 - 2 | 923 | 75 | |
BT005964.1 | - | 1 - 2 | 784 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |