Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCHX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100690.3 | NP_172294.2 | 2 - 3 | 1474 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |