Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100720.3 | NP_563818.1 | 3 - 4 | 970 | 85 | |
GenBank mRNA | AF419594.1 | - | 3 - 4 | 887 | 85 | |
AY064047.1 | - | 3 - 4 | 817 | 85 | ||
AY117314.1 | - | 3 - 4 | 776 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |