Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FATB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100724.3 | NP_172327.1 | 2 - 3 | 1014 | 76 | |
GenBank mRNA | AY054679.1 | - | 2 - 3 | 837 | 76 | |
AY059827.1 | - | 2 - 3 | 638 | 76 | ||
AY064652.1 | - | 1 - 2 | 496 | 76 | ||
BT008505.1 | - | 1 - 2 | 496 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |