Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100727.4 | NP_172330.1 | 1 - 2 | 247 | 80 | |
GenBank mRNA | AB004293.1 | - | 1 - 2 | 203 | 80 | |
AB004820.1 | - | 1 - 2 | 168 | 80 | ||
BT030346.1 | - | 1 - 2 | 81 | 80 | ||
Y15362.1 | - | 132 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |