Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100727.4 | NP_172330.1 | 2 - 3 | 431 | 69 | |
GenBank mRNA | AB004293.1 | - | 2 - 3 | 387 | 69 | |
AB004820.1 | - | 2 - 3 | 352 | 69 | ||
BT030346.1 | - | 2 - 3 | 265 | 69 | ||
Y15362.1 | - | 316 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |