Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035923.3 | NP_001031000.1 | 2 - 3 | 446 | 123 | |
NM_001331777.1 | NP_001321301.1 | 1 - 2 | 809 | 123 | ||
NM_100728.4 | NP_172331.1 | 2 - 3 | 514 | 123 | ||
GenBank mRNA | AF370251.1 | - | 2 - 3 | 370 | 123 | |
AK317253.1 | - | 2 - 3 | 289 | 123 | ||
AY063067.1 | - | 1 - 2 | 215 | 123 | ||
U44133.1 | - | 1 - 2 | 259 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |