Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035923.3 | NP_001031000.1 | 5 - 6 | 1107 | 122 | |
NM_001331777.1 | NP_001321301.1 | 4 - 5 | 1470 | 122 | ||
NM_100728.4 | NP_172331.1 | 5 - 6 | 1175 | 122 | ||
GenBank mRNA | AF370251.1 | - | 5 - 6 | 1031 | 122 | |
AK317253.1 | - | 5 - 6 | 950 | 122 | ||
AY063067.1 | - | 4 - 5 | 876 | 122 | ||
U44133.1 | - | 4 - 5 | 920 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |